Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003906.4 | NP_003897.2 | 5 | 1799 | ||
XM_005261203.4 | XP_005261260.1 | 6 | 2375 | |||
XM_005261204.4 | XP_005261261.1 | 6 | 2271 | |||
XM_005261205.3 | XP_005261262.1 | 6 | 1887 | |||
GenBank mRNA | AB011144.1 | - | 5 | 1369 | ||
AJ010089.1 | - | 5 | 1728 | |||
AK304208.1 | - | 5 | 658 | |||
BC104958.1 | - | 5 | 1767 | 8143961 | ||
BC104960.1 | - | 5 | 1767 | 8143963 |