Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003906.4 | NP_003897.2 | 22, 23 | 4752 | ||
XM_005261203.4 | XP_005261260.1 | 23, 24 | 5328 | |||
XM_005261204.4 | XP_005261261.1 | 23, 24 | 5224 | |||
XM_005261205.3 | XP_005261262.1 | 23, 24 | 4840 | |||
GenBank mRNA | AB005543.1 | - | 7, 8 | 1237 | ||
AB011144.1 | - | 22, 23 | 4322 | |||
AJ010089.1 | - | 22, 23 | 4681 | |||
AK026578.1 | - | 1, 2 | 95 | |||
AK095181.1 | - | 2, 3 | 1705 | |||
AK126313.1 | - | 12, 13 | 3157 | |||
AK127239.1 | - | 12, 13 | 2641 | |||
AK304208.1 | - | 22, 23 | 3611 | |||
BC004497.2 | - | 3, 4 | 402 | |||
BC013285.2 | - | 7, 8 | 1038 | |||
BC104958.1 | - | 22, 23 | 4720 | 8143961 | ||
BC104960.1 | - | 22, 23 | 4720 | 8143963 |