Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003909.4 | NP_003900.1 | 5 | 501 | ||
XM_005251093.4 | XP_005251150.1 | 4 | 453 | |||
XM_017013945.1 | XP_016869434.1 | 6 | 626 | |||
GenBank mRNA | AB014536.1 | - | 5 | 458 | ||
AF077226.2 | - | 4 | 399 | |||
AK292912.1 | - | 5 | 490 | |||
BC007017.1 | - | 5 | 485 | |||
BC008385.1 | - | 5 | 485 | |||
BC036242.1 | - | 5 | 461 | 5300530 | ||
BC066597.1 | - | 5 | 472 | 6168840 | ||
DA780310.1 | - | 5 | 502 | |||
DB135485.1 | - | 5 | 501 |