Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003909.4 | NP_003900.1 | 16 | 1481 | ||
XM_005251093.4 | XP_005251150.1 | 15 | 1433 | |||
XM_017013945.1 | XP_016869434.1 | 17 | 1606 | |||
GenBank mRNA | AB014536.1 | - | 16 | 1438 | ||
AF077226.2 | - | 15 | 1379 | |||
AK124311.1 | - | 1 | 634 | |||
AK292912.1 | - | 16 | 1470 | |||
BC036242.1 | - | 16 | 1441 | 5300530 | ||
BC066597.1 | - | 16 | 1452 | 6168840 | ||
BX647342.1 | - | 6 | 904 |