Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272071.1 | NP_001259000.1 | 6 | 2022 | ||
NM_003916.4 | NP_003907.3 | 5 | 2013 | |||
GenBank mRNA | AK125525.1 | - | 1 | 3164 | ||
AK130075.1 | - | 5 | 1816 | |||
BX537780.1 | - | 5 | 1707 |