Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261826.1 | NP_001248755.1 | 11 | 1157 | ||
NM_003938.6 | NP_003929.4 | 11 | 1157 | |||
XM_006722932.2 | XP_006722995.1 | 11 | 1220 | |||
XM_017027422.1 | XP_016882911.1 | 6 | 454 | |||
GenBank mRNA | AB208804.1 | - | 11 | 1141 | ||
AF002163.1 | - | 1245 | ||||
AF130042.1 | - | 9 | 716 | |||
BC005142.2 | - | 11 | 985 | |||
BC010065.2 | - | 11 | 1136 | 4106961 | ||
U91930.1 | - | 11 | 870 |