Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003958.3 | NP_003949.1 | 3 | 488 | ||
NM_183078.2 | NP_898901.1 | 3 | 488 | |||
NR_046399.1 | 3 | 594 | ||||
XM_006715241.2 | XP_006715304.1 | 3 | 488 | |||
XM_006715242.2 | XP_006715305.1 | 3 | 488 | |||
XM_017011462.1 | XP_016866951.1 | 3 | 339 | |||
XM_017011463.1 | XP_016866952.1 | 3 | 344 | |||
XM_017011464.1 | XP_016866953.1 | 3 | 493 | |||
GenBank mRNA | AB014546.1 | - | 3 | 407 | ||
AF334675.1 | - | 3 | 295 | |||
AK022075.1 | - | 3 | 389 | |||
AK222765.1 | - | 3 | 406 | |||
AK298319.1 | - | 3 | 512 | |||
AK308840.1 | - | 3 | 395 | |||
AL711279.1 | - | 3 | 446 | |||
BC007517.2 | - | 3 | 417 | 3028746 | ||
BT007446.1 | - | 3 | 295 | |||
DA064222.1 | - | 3 | 488 |