Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130910.1 | NP_001124382.1 | 2 | 1446 | ||
NM_003965.4 | NP_003956.2 | 2 | 1582 | |||
XM_011534208.1 | XP_011532510.1 | 3 | 1526 | |||
XM_011534209.1 | XP_011532511.1 | 3 | 1449 | |||
XM_017007436.1 | XP_016862925.1 | 2 | 1442 | |||
GenBank mRNA | AF015524.1 | - | 2 | 1380 | ||
AF015525.1 | - | 2 | 1624 | |||
BC071682.1 | - | 2 | 1535 | 5182006 | ||
BC096075.1 | - | 2 | 1362 | 40000578 | ||
BC096076.3 | - | 2 | 1362 | 40000580 |