Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034996.2 | NP_001030168.1 | 6 | 885 | ||
NM_003973.4 | NP_003964.3 | 6 | 821 | |||
GenBank mRNA | AK290674.1 | - | 6 | 784 | ||
BC005134.2 | - | 5 | 773 | 3928410 |