Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004227.3 | NP_004218.1 | 13 | 1918 | ||
GenBank mRNA | AK023609.1 | - | 1 | 41 | ||
BC008191.1 | - | 6 | 1157 | |||
BC028717.1 | - | 13 | 1954 | 4811560 |