Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288962.1 | NP_001275891.1 | 6 | 502 | ||
NM_001288963.1 | NP_001275892.1 | 6 | 502 | |||
NM_004240.3 | NP_004231.1 | 6 | 502 | |||
NR_110231.1 | 6 | 502 | ||||
XM_005259683.2 | XP_005259740.1 | 6 | 502 | |||
XM_006722940.1 | XP_006723003.1 | 6 | 502 | |||
GenBank mRNA | AB007845.1 | - | 6 | 477 | ||
AB072596.1 | - | 6 | 477 | |||
AF380114.1 | - | 6 | 422 | |||
AF502289.1 | - | 6 | 461 | |||
AJ000414.1 | - | 6 | 461 | |||
AK223109.1 | - | 6 | 467 | |||
AK313296.1 | - | 6 | 484 | |||
AL832186.1 | - | 6 | 588 | |||
AY081141.1 | - | 6 | 456 | |||
BC013002.2 | - | 6 | 447 | 3532036 | ||
BT006698.1 | - | 6 | 422 | |||
BT020167.1 | - | 6 | 422 | |||
BT020171.1 | - | 6 | 422 | |||
CB997985.1 | - | 6 | 515 | |||
CR536513.1 | - | 6 | 422 | |||
DB198268.1 | - | 6 | 504 |