Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277077.1 | NP_001264006.1 | 6 | 2917 | ||
NM_001277078.1 | NP_001264007.1 | 6 | 2958 | |||
NM_004272.4 | NP_004263.1 | 9 | 3307 | |||
GenBank mRNA | BC047438.1 | - | 9 | 3307 | 5294849 |