Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004288.4 | NP_004279.3 | 1 | 178 | ||
GenBank mRNA | AK312753.1 | - | 1 | 175 | ||
BC036449.2 | - | 1 | 136 | 4830272 | ||
CD642163.1 | - | 1 | 231 | |||
DB219691.1 | - | 1 | 179 |