Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324423.1 | NP_001311352.1 | 3 | 397 | ||
NM_004664.3 | NP_004655.1 | 4 | 516 | |||
NR_136887.1 | 4 | 474 | ||||
NR_136888.1 | 5 | 593 | ||||
XM_011538928.2 | XP_011537230.1 | 4 | 510 | |||
GenBank mRNA | AF028826.1 | - | 3 | 232 | ||
AF087693.1 | - | 4 | 485 | |||
AF173081.1 | - | 4 | 516 | |||
AK309256.1 | - | 4 | 305 | |||
AK315321.1 | - | 4 | 360 | |||
BC062332.1 | - | 5 | 458 | |||
BC099921.1 | - | 4 | 326 | 6729473 | ||
BC118609.1 | - | 4 | 460 | 40109202 | ||
BC122561.1 | - | 4 | 460 | 40109204 | ||
CR407680.1 | - | 4 | 323 |