Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004714.2 | NP_004705.1 | 5 | 820 | ||
NM_006483.2 | NP_006474.1 | 5 | 820 | |||
NM_006484.2 | NP_006475.1 | 5 | 820 | |||
XM_005259398.4 | XP_005259455.1 | 5 | 1272 | |||
GenBank mRNA | AF205861.1 | - | 5 | 717 | ||
AK074971.1 | - | 5 | 736 | |||
AK302127.1 | - | 5 | 749 | |||
BC018751.1 | - | 5 | 772 | 4300012 | ||
BC025291.1 | - | 5 | 723 | 5019417 | ||
Y17999.1 | - | 5 | 788 |