Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004724.3 | NP_004715.1 | 3 | 396 | ||
XM_017018558.1 | XP_016874047.1 | 3 | 336 | |||
XM_017018559.1 | XP_016874048.1 | 3 | 336 | |||
GenBank mRNA | AK300746.1 | - | 3 | 354 | ||
AK303871.1 | - | 2 | 75 | |||
AK312925.1 | - | 3 | 374 | |||
BC128264.1 | - | 3 | 324 | 40118818 | ||
BM833775.1 | - | 3 | 396 | |||
U54996.1 | - | 3 | 322 |