Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195677.1 | NP_001182606.1 | 3 | 969 | ||
NM_004738.4 | NP_004729.1 | 6 | 1331 | |||
NR_036633.1 | 4 | 1146 | ||||
GenBank mRNA | AB209665.1 | - | 2 | 584 | ||
AF086628.1 | - | 6 | 1137 | |||
AF086629.1 | - | 3 | 775 | |||
AF160212.1 | - | 6 | 1029 | |||
AK023241.1 | - | 6 | 1328 | |||
AK024771.1 | - | 1 | 237 | |||
AK123538.1 | - | 6 | 1334 | |||
AK127252.1 | - | 6 | 1184 | |||
AK128422.1 | - | 2 | 2525 | |||
AK298036.1 | - | 4 | 899 | |||
AL833440.1 | - | 4 | 720 | |||
AY358464.1 | - | 6 | 1224 | |||
BC001712.1 | - | 7 | 1168 | 3543354 |