Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003809.2 | NP_001003809.1 | 4, 5 | 767 | ||
NM_001242761.1 | NP_001229690.1 | 7, 8 | 2124 | |||
NM_001242764.1 | NP_001229693.1 | 4, 5 | 740 | |||
NM_001242765.1 | NP_001229694.1 | 4, 5 | 767 | |||
NM_004746.3 | NP_004737.2 | 7, 8 | 2124 | |||
XM_017026080.1 | XP_016881569.1 | 5, 6 | 1123 | |||
XM_017026081.1 | XP_016881570.1 | 4, 5 | 787 | |||
XM_017026082.1 | XP_016881571.1 | 5, 6 | 942 | |||
XM_017026083.1 | XP_016881572.1 | 4, 5 | 1012 | |||
XM_017026084.1 | XP_016881573.1 | 4, 5 | 1009 | |||
XM_017026085.1 | XP_016881574.1 | 4, 5 | 1002 | |||
GenBank mRNA | AB000276.1 | - | 4, 5 | 710 | ||
AB000277.1 | - | 4, 5 | 1664 | |||
AK299880.1 | - | 4, 5 | 740 | |||
AK304101.1 | - | 7, 8 | 2106 | |||
AK316099.1 | - | 7, 8 | 2124 | |||
BC136453.1 | - | 4, 5 | 1656 | 9020442 | ||
BC136454.1 | - | 4, 5 | 1656 | 9020443 | ||
U67988.1 | - | 4, 5 | 714 |