Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289979.1 | NP_001276908.1 | 9 | 2148 | ||
NM_001289980.1 | NP_001276909.1 | 9 | 2032 | |||
NM_004756.4 | NP_004747.1 | 10 | 2233 | |||
GenBank mRNA | AF015041.1 | - | 10 | 2267 | ||
AK289398.1 | - | 10 | 2219 | |||
BC001794.1 | - | 9 | 2121 | |||
BC144583.1 | - | 9 | 2037 |