Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261464.1 | NP_001248393.1 | 16 | 2118 | ||
NM_001261465.1 | NP_001248394.1 | 16 | 2088 | |||
NM_003815.4 | NP_003806.3 | 16 | 2088 | |||
NM_207191.2 | NP_997074.1 | 16 | 2088 | |||
NM_207194.2 | NP_997077.1 | 16 | 2088 | |||
NM_207195.2 | NP_997078.1 | 16 | 2088 | |||
NM_207196.2 | NP_997079.1 | 16 | 2088 | |||
NM_207197.2 | NP_997080.1 | 16 | 2088 | |||
NR_048577.1 | 16 | 2088 | ||||
NR_048578.1 | 16 | 2088 | ||||
NR_048579.1 | 16 | 2088 | ||||
GenBank mRNA | AB209157.1 | - | 15 | 3619 | ||
AK075498.1 | - | 8 | 1162 | |||
AK295614.1 | - | 16 | 2060 | |||
AK296925.1 | - | 16 | 2039 | |||
AY518542.1 | - | 16 | 1973 | |||
AY560593.1 | - | 16 | 2074 | |||
AY560594.1 | - | 16 | 2074 | |||
AY560595.1 | - | 16 | 2074 | |||
AY560596.1 | - | 16 | 2074 | |||
AY560597.1 | - | 16 | 2074 | |||
AY560598.1 | - | 16 | 2074 | |||
AY560599.1 | - | 16 | 2074 | |||
AY560600.1 | - | 16 | 2074 | |||
AY560601.1 | - | 16 | 2074 | |||
AY576417.1 | - | 16 | 2074 | |||
BC014566.2 | - | 16 | 2054 | 2958341 | ||
BT009764.1 | - | 16 | 1973 | |||
U41767.1 | - | 16 | 1980 | |||
U46005.1 | - | 16 | 1973 |