Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 14 | 2078 | ||
NM_001292019.1 | NP_001278948.1 | 14 | 1644 | |||
NM_004762.3 | NP_004753.1 | 14 | 1681 | |||
NM_017456.3 | NP_059430.2 | 13 | 1678 | |||
XM_011525475.2 | XP_011523777.1 | 13 | 2421 | |||
XM_011525476.2 | XP_011523778.1 | 14 | 1596 | |||
XM_011525477.1 | XP_011523779.1 | 12 | 1459 | |||
XM_011525478.1 | XP_011523780.1 | 13 | 2000 | |||
XM_017025336.1 | XP_016880825.1 | 13 | 1748 | |||
XM_017025337.1 | XP_016880826.1 | 14 | 1689 | |||
XM_017025338.1 | XP_016880827.1 | 13 | 2064 | |||
GenBank mRNA | AF054997.1 | - | 1 | 41 | ||
AK091369.1 | - | 1 | 1180 | |||
AK123894.1 | - | 1 | 2745 | |||
AL832791.1 | - | 1 | 1859 | |||
BC038385.1 | - | 14 | 1602 | 4755203 | ||
BC050452.1 | - | 14 | 1591 | 6043039 | ||
M85169.1 | - | 14 | 1631 |