Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004800.2 | NP_004791.1 | 1, 2 | 403 | ||
GenBank mRNA | AK225800.1 | - | 1, 2 | 303 | ||
AK290514.1 | - | 1, 2 | 358 | |||
AK315210.1 | - | 1, 2 | 358 | |||
BC110656.1 | - | 1, 2 | 360 | 3855486 | ||
BC130293.1 | - | 1, 2 | 237 | 40146324 | ||
DB024878.1 | - | 1, 2 | 403 | |||
U81006.1 | - | 1, 2 | 303 |