Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM189A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127608.1 | NP_001121080.1 | 6 | 573 | ||
NM_004816.3 | NP_004807.3 | 6 | 889 | |||
XM_005252307.4 | XP_005252364.1 | 6 | 1450 | |||
XM_017015324.1 | XP_016870813.1 | 5 | 634 | |||
XM_017015325.1 | XP_016870814.1 | 6 | 573 | |||
GenBank mRNA | AK056152.1 | - | 4 | 517 | ||
AK126127.1 | - | 6 | 889 | |||
BC021685.1 | - | 3 | 302 | |||
BC113683.1 | - | 6 | 532 | 8322735 | ||
BC113685.1 | - | 6 | 532 | 8322737 | ||
BX641153.1 | - | 3 | 5742 |