Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039477.2 | NP_001034566.1 | 3 | 331 | ||
NM_001105556.2 | NP_001099026.1 | 5 | 1815 | |||
NM_001286113.1 | NP_001273042.1 | 6 | 1428 | |||
NM_001286115.1 | NP_001273044.1 | 5 | 1227 | |||
NM_004848.3 | NP_004839.2 | 4 | 742 | |||
XM_005246041.3 | XP_005246098.1 | 4 | 1416 | |||
XM_006711050.1 | XP_006711113.1 | 6 | 1374 | |||
XM_011542445.2 | XP_011540747.1 | 4 | 1686 | |||
GenBank mRNA | AB035482.1 | - | 2 | 287 | ||
AB050854.1 | - | 4 | 1032 | |||
AF044896.1 | - | 3 | 1469 | |||
AK094833.1 | - | 1 | 1964 | |||
AK303090.1 | - | 6 | 1390 | |||
AK303141.1 | - | 5 | 1223 | |||
BC031655.1 | - | 4 | 710 | |||
BC132692.1 | - | 4 | 741 | |||
BC133049.1 | - | 4 | 741 | |||
BG716895.1 | - | 3 | 324 | |||
BI908174.1 | - | 2 | 337 | |||
CR749321.1 | - | 3 | 332 |