Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004870.3 | NP_004861.2 | 7 | 974 | ||
NR_024603.1 | 7 | 969 | ||||
XM_006721597.2 | XP_006721660.1 | 6 | 654 | |||
XM_006721598.2 | XP_006721661.1 | 6 | 916 | |||
XM_006721599.2 | XP_006721662.1 | 6 | 715 | |||
XM_011524081.2 | XP_011522383.1 | 7 | 805 | |||
GenBank mRNA | AF038961.1 | - | 7 | 774 | ||
AF059752.1 | - | 7 | 813 | |||
AF258568.1 | - | 2 | 943 | |||
AK055923.1 | - | 7 | 689 | |||
AK075299.1 | - | 7 | 773 | |||
AK225401.1 | - | 7 | 773 | |||
AK225432.1 | - | 7 | 769 | |||
AK297001.1 | - | 6 | 912 | |||
AK300140.1 | - | 7 | 760 | |||
BC001898.1 | - | 7 | 785 | 3536216 | ||
BG776573.1 | - | 3 | 313 | |||
DQ499597.1 | - | 7 | 756 |