Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305043.1 | NP_001291972.1 | 3 | 585 | ||
NM_001305049.1 | NP_001291978.1 | 3 | 364 | |||
NM_001305051.1 | NP_001291980.1 | 3 | 364 | |||
NM_001305052.1 | NP_001291981.1 | 2 | 258 | |||
NM_001305066.1 | NP_001291995.1 | 3 | 585 | |||
NM_004872.4 | NP_004863.2 | 3 | 585 | |||
GenBank mRNA | AF047439.1 | - | 3 | 349 | ||
AF290615.1 | - | 3 | 585 | |||
AK075187.1 | - | 3 | 525 | |||
AK075505.1 | - | 3 | 348 | |||
AK123445.1 | - | 2 | 788 | |||
AK225624.1 | - | 3 | 350 | |||
AX775907.1 | - | 3 | 585 | |||
AX775909.1 | - | 3 | 352 | |||
AY359029.1 | - | 3 | 404 | |||
BC003106.1 | - | 3 | 334 | 3501639 | ||
BC016374.1 | - | 3 | 352 | 4618034 | ||
BC062738.1 | - | 2 | 83 | |||
BC095464.1 | - | 3 | 377 | 30554552 | ||
BF574560.1 | - | 2 | 98 | |||
BP371330.1 | - | 3 | 363 | |||
BU165003.1 | - | 3 | 223 | |||
BX463615.2 | - | 3 | 222 | |||
DB459209.1 | - | 3 | 363 |