Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305043.1 | NP_001291972.1 | 4 | 772 | ||
NM_001305049.1 | NP_001291978.1 | 4 | 551 | |||
NM_001305050.1 | NP_001291979.1 | 2 | 571 | |||
NM_001305051.1 | NP_001291980.1 | 4 | 551 | |||
NM_001305052.1 | NP_001291981.1 | 3 | 445 | |||
NM_004872.4 | NP_004863.2 | 4 | 772 | |||
XM_006711051.1 | XP_006711114.1 | 2 | 569 | |||
GenBank mRNA | AF047439.1 | - | 4 | 536 | ||
AF290615.1 | - | 4 | 772 | |||
AK075187.1 | - | 4 | 712 | |||
AK075505.1 | - | 4 | 535 | |||
AK123445.1 | - | 3 | 975 | |||
AK124967.1 | - | 1 | 1557 | |||
AK225624.1 | - | 4 | 537 | |||
AX775907.1 | - | 4 | 772 | |||
AX775909.1 | - | 4 | 539 | |||
AY359029.1 | - | 4 | 591 | |||
BC003106.1 | - | 4 | 521 | 3501639 | ||
BC016374.1 | - | 4 | 539 | 4618034 | ||
BC062738.1 | - | 3 | 270 | |||
BC095464.1 | - | 4 | 564 | 30554552 | ||
BF574560.1 | - | 3 | 285 | |||
BI604139.1 | - | 2 | 339 | |||
BP371330.1 | - | 4 | 550 | |||
BU165003.1 | - | 4 | 410 | |||
BX463615.2 | - | 4 | 409 |