Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301008.1 | NP_001287937.1 | 5, 6 | 378 | ||
NM_004877.3 | NP_004868.1 | 6, 7 | 475 | |||
GenBank mRNA | AB001993.1 | - | 5, 6 | 375 | ||
AF038956.1 | - | 5, 6 | 369 | |||
BC022207.1 | - | 6, 7 | 395 | |||
BC032819.2 | - | 5, 6 | 362 | |||
BC080180.1 | - | 6, 7 | 395 | 5419511 | ||
BC093799.1 | - | 6, 7 | 428 | 7939644 | ||
BC101818.1 | - | 6, 7 | 428 | 8069324 | ||
BC143543.1 | - | 6, 7 | 428 | 9052054 | ||
BC143546.1 | - | 6, 7 | 427 | 9052057 | ||
BC143548.1 | - | 6, 7 | 426 | 9052059 | ||
BF974742.1 | - | 6, 7 | 476 | |||
CR456967.1 | - | 5, 6 | 365 | |||
CR542114.1 | - | 5, 6 | 365 | |||
EB387540.1 | - | 6, 7 | 574 |