Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 11 | 2035 | ||
NM_004879.4 | NP_004870.3 | 11 | 2077 | |||
NR_110769.1 | 10 | 1931 | ||||
NR_110770.1 | 12 | 2202 | ||||
XM_011543069.1 | XP_011541371.1 | 12 | 2093 | |||
XM_011543070.1 | XP_011541372.1 | 10 | 2136 | |||
XM_017018582.1 | XP_016874071.1 | 11 | 1981 | |||
GenBank mRNA | AF010313.2 | - | 11 | 1949 | ||
BQ936285.1 | - | 1 | 213 | |||
DB520736.1 | - | 1 | 225 | |||
DB567734.1 | - | 1 | 266 |