Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGDCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004884.3 | NP_004875.2 | 2, 3 | 670 | ||
XM_011522241.1 | XP_011520543.1 | 2, 3 | 612 | |||
XM_011522245.2 | XP_011520547.1 | 2, 3 | 612 | |||
GenBank mRNA | BC042054.1 | - | 2, 3 | 618 | 5579570 | |
BC067107.1 | - | 2, 3 | 670 | 6644528 |