Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004890.2 | NP_004881.2 | 3, 4 | 274 | ||
XM_006721600.2 | XP_006721663.1 | 3, 4 | 899 | |||
GenBank mRNA | AF047437.1 | - | 3, 4 | 251 | ||
AK123065.1 | - | 1053 | ||||
AK312174.1 | - | 3, 4 | 250 | |||
BC011934.2 | - | 3, 4 | 243 | 4330461 | ||
BP308951.1 | - | 3, 4 | 273 |