Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MINPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178117.1 | NP_001171588.1 | 3 | 1400 | ||
NM_001178118.1 | NP_001171589.1 | 5 | 951 | |||
NM_004897.4 | NP_004888.2 | 5 | 1632 | |||
XM_006718078.2 | XP_006718141.1 | 4 | 3172 | |||
XM_011540379.2 | XP_011538681.1 | 6 | 1318 | |||
XM_017016966.1 | XP_016872455.1 | 6 | 1337 | |||
GenBank mRNA | AB209819.1 | - | 4 | 1097 | ||
AF046914.1 | - | 5 | 1182 | |||
AF084943.1 | - | 5 | 1202 | |||
AF084944.1 | - | 3 | 955 | |||
AK304624.1 | - | 6 | 823 | |||
AK309176.1 | - | 5 | 950 | |||
AK312937.1 | - | 5 | 1223 | |||
AL050356.1 | - | 5 | 1205 | |||
AY358938.1 | - | 5 | 1187 | |||
BC032504.1 | - | 5 | 1184 | 5243440 |