Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270531.1 | NP_001257460.1 | 6 | 562 | ||
NM_001270532.1 | NP_001257461.1 | 6 | 694 | |||
NM_001270533.1 | NP_001257462.1 | 6 | 562 | |||
NM_004906.4 | NP_004897.2 | 6 | 694 | |||
NM_152857.2 | NP_690596.1 | 6 | 562 | |||
NM_152858.2 | NP_690597.1 | 6 | 694 | |||
XM_017011514.1 | XP_016867003.1 | 6 | 532 | |||
XM_017011515.1 | XP_016867004.1 | 4 | 794 | |||
XM_017011516.1 | XP_016867005.1 | 6 | 532 | |||
GenBank mRNA | AF277190.1 | - | 2 | 134 | ||
AF374416.1 | - | 6 | 575 | |||
AJ276706.1 | - | 5 | 342 | |||
AK127822.1 | - | 6 | 573 | |||
AK290854.1 | - | 6 | 575 | |||
AL583911.1 | - | 1 | 19 | |||
BC000383.2 | - | 6 | 553 | |||
BC004432.1 | - | 6 | 553 | |||
BC069192.1 | - | 6 | 530 | 4653831 | ||
BG702882.1 | - | 6 | 566 | |||
BQ948913.1 | - | 6 | 500 | |||
BX387163.2 | - | 6 | 544 | |||
CN284477.1 | - | 6 | 524 | |||
D14661.1 | - | 6 | 474 |