Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004913.2 | NP_004904.2 | 15 | 2726 | ||
XM_005256329.4 | XP_005256386.1 | 15 | 2789 | |||
XM_005256330.4 | XP_005256387.1 | 15 | 2606 | |||
XM_005256331.2 | XP_005256388.1 | 12 | 2263 | |||
XM_006721350.2 | XP_006721413.1 | 14 | 2684 | |||
XM_011523476.2 | XP_011521778.1 | 15 | 2786 | |||
XM_011523477.2 | XP_011521779.1 | 14 | 4602 | |||
XM_011523478.2 | XP_011521780.1 | 14 | 4599 | |||
XM_011523480.2 | XP_011521782.1 | 15 | 2643 | |||
GenBank mRNA | AB018551.1 | - | 14 | 2763 |