Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004918.3 | NP_004909.1 | 4 | 1097 | ||
GenBank mRNA | AB018563.1 | - | 3 | 1668 | ||
AF110466.1 | - | 4 | 1073 | |||
AF137027.1 | - | 4 | 1092 | |||
BC041616.1 | - | 4 | 1071 | 5816694 | ||
BC051000.1 | - | 4 | 1097 | 5813497 |