Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039656.1 | NP_001034745.1 | 2 | 289 | ||
NM_004923.3 | NP_004914.2 | 2 | 289 | |||
XM_011545402.1 | XP_011543704.1 | 2 | 333 | |||
XM_011545403.2 | XP_011543705.1 | 1 | 202 | |||
XM_017018588.1 | XP_016874077.1 | 2 | 333 | |||
GenBank mRNA | AK074817.1 | - | 2 | 236 | ||
AK292358.1 | - | 2 | 229 | |||
BC027593.1 | - | 2 | 225 | 4838081 | ||
BC054047.2 | - | 1 | 100 | |||
BC064579.1 | - | 2 | 256 | 5743865 | ||
BG772890.1 | - | 2 | 307 | |||
BM790577.1 | - | 2 | 289 |