Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005095.2 | NP_005086.2 | 3 | 214 | ||
XM_005271328.4 | XP_005271385.1 | 3 | 493 | |||
XM_011542421.2 | XP_011540723.1 | 3 | 140 | |||
XM_011542423.2 | XP_011540725.1 | 4 | 545 | |||
XM_017002803.1 | XP_016858292.1 | 4 | 267 | |||
XM_017002804.1 | XP_016858293.1 | 3 | 131 | |||
XM_017002805.1 | XP_016858294.1 | 1 | 46 | |||
GenBank mRNA | AK308320.1 | - | 3 | 214 | ||
BC127113.1 | - | 4 | 220 | 40125110 | ||
BG499626.1 | - | 3 | 215 | |||
CD653525.1 | - | 2 | 108 | |||
CR936741.1 | - | 2 | 78 |