Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003849.3 | NP_003840.2 | 2 | 384 | ||
GenBank mRNA | AF104921.2 | - | 2 | 183 | ||
AK225737.1 | - | 2 | 161 | |||
AK291082.1 | - | 2 | 183 | |||
AK296741.1 | - | 2 | 191 | |||
AK313290.1 | - | 2 | 163 | |||
BC000504.2 | - | 2 | 187 | 2821634 | ||
BM804835.1 | - | 2 | 370 | |||
BP194715.1 | - | 2 | 385 | |||
CR457139.1 | - | 2 | 152 | |||
Z68204.1 | - | 2 | 158 |