Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005127.2 | NP_005118.2 | 5 | 1065 | ||
GenBank mRNA | AB015628.1 | - | 5 | 558 | ||
AK313916.1 | - | 5 | 569 | |||
BC005254.1 | - | 5 | 559 | 3996700 | ||
BG540007.1 | - | 1 | 304 | |||
CB160326.1 | - | 3 | 288 | |||
X96719.1 | - | 5 | 540 |