Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOPEY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320714.1 | NP_001307643.1 | 3 | 443 | ||
NM_005128.3 | NP_005119.2 | 3 | 334 | |||
GenBank mRNA | AJ237839.2 | - | 3 | 340 | ||
BP364624.1 | - | 3 | 401 | |||
BP365117.1 | - | 3 | 443 |