Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135110.2 | NP_001128582.1 | 5 | 832 | ||
NM_001286516.1 | NP_001273445.1 | 3 | 483 | |||
NM_005147.5 | NP_005138.3 | 5 | 832 | |||
GenBank mRNA | AB209923.1 | - | 5 | 751 | ||
AF061749.1 | - | 5 | 786 | |||
AF244136.1 | - | 5 | 757 | |||
AF411044.1 | - | 5 | 799 | |||
AK001333.1 | - | 5 | 772 | |||
AK223105.1 | - | 5 | 772 | |||
AK295391.1 | - | 3 | 423 | |||
AK314218.1 | - | 5 | 769 | |||
BC007225.1 | - | 5 | 752 | |||
BC011855.2 | - | 5 | 756 | 4139881 | ||
BC012343.1 | - | 5 | 748 | 4653507 | ||
BC014062.2 | - | 5 | 748 | |||
BC020248.1 | - | 1 | 81 | |||
BC030145.2 | - | 5 | 748 | |||
BC032100.2 | - | 5 | 770 | 4660602 | ||
BX445320.2 | - | 5 | 832 |