Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135110.2 | NP_001128582.1 | 11 | 1710 | ||
NM_001286516.1 | NP_001273445.1 | 9 | 1361 | |||
NM_005147.5 | NP_005138.3 | 12 | 1827 | |||
GenBank mRNA | AB209923.1 | - | 12 | 1746 | ||
AF244136.1 | - | 11 | 1638 | |||
AF411044.1 | - | 11 | 1677 | |||
AK001333.1 | - | 11 | 1650 | |||
AK127660.1 | - | 3 | 1355 | |||
AK223105.1 | - | 12 | 1767 | |||
AK295391.1 | - | 9 | 1301 | |||
BC007225.1 | - | 12 | 1747 | |||
BC011855.2 | - | 12 | 1751 | 4139881 | ||
BC012343.1 | - | 11 | 1626 | 4653507 | ||
BC014062.2 | - | 11 | 1626 | |||
BC020248.1 | - | 7 | 959 | |||
BC030145.2 | - | 11 | 1626 | |||
BC032100.2 | - | 11 | 1648 | 4660602 |