Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271634.1 | NP_001258563.1 | 4 | 771 | ||
NM_001271635.1 | NP_001258564.1 | 4 | 771 | |||
NM_005444.2 | NP_005435.1 | 4 | 771 | |||
XM_011512138.2 | XP_011510440.1 | 5 | 890 | |||
XM_017005248.1 | XP_016860737.1 | 4 | 1360 | |||
XM_017005249.1 | XP_016860738.1 | 5 | 892 | |||
GenBank mRNA | AB500892.1 | - | 4 | 643 | ||
AK293281.1 | - | 4 | 771 | |||
AK316567.1 | - | 4 | 477 | |||
BC007102.1 | - | 4 | 478 | |||
BC137455.1 | - | 4 | 493 | 9021455 | ||
BC137456.1 | - | 4 | 493 | 9021456 |