Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206729.1 | NP_001193658.1 | 9 | 899 | ||
NM_005465.4 | NP_005456.1 | 8 | 858 | |||
NM_181690.2 | NP_859029.1 | 8 | 858 | |||
XM_005272994.4 | XP_005273051.1 | 9 | 1083 | |||
XM_005272995.2 | XP_005273052.1 | 9 | 889 | |||
XM_011544012.2 | XP_011542314.1 | 9 | 1083 | |||
XM_011544013.2 | XP_011542315.1 | 9 | 1083 | |||
XM_016999985.1 | XP_016855474.1 | 10 | 1031 | |||
GenBank mRNA | AF085234.1 | - | 8 | 793 | ||
AF124141.1 | - | 8 | 758 | |||
AF135794.1 | - | 8 | 746 | |||
AJ245709.1 | - | 8 | 756 | |||
AK308052.1 | - | 8 | 784 | |||
AL117525.1 | - | 9 | 899 | |||
AY005799.1 | - | 8 | 819 | |||
BC020479.1 | - | 8 | 792 | |||
BC121154.2 | - | 8 | 829 | 40121374 |