Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012750.2 | NP_001012768.1 | 8 | 1098 | ||
NM_001012751.2 | NP_001012769.1 | 8 | 1098 | |||
NM_001012752.2 | NP_001012770.1 | 7 | 1083 | |||
NM_001178116.1 | NP_001171587.1 | 10 | 1230 | |||
NM_001178119.1 | NP_001171590.1 | 8 | 1095 | |||
NM_001178120.1 | NP_001171591.1 | 8 | 1095 | |||
NM_001178121.1 | NP_001171592.1 | 7 | 1080 | |||
NM_005470.3 | NP_005461.2 | 9 | 1179 | |||
XM_005252332.3 | XP_005252389.1 | 8 | 979 | |||
XM_005252333.3 | XP_005252390.1 | 7 | 895 | |||
XM_006717359.2 | XP_006717422.1 | 9 | 1075 | |||
XM_006717360.2 | XP_006717423.1 | 9 | 1078 | |||
XM_006717361.2 | XP_006717424.1 | 7 | 898 | |||
XM_017015458.1 | XP_016870947.1 | 9 | 1075 | |||
GenBank mRNA | AB040151.1 | - | 8 | 1041 | ||
AB209268.1 | - | 8 | 946 | |||
AF001628.1 | - | 8 | 919 | |||
AF006516.1 | - | 8 | 937 | |||
AF260262.1 | - | 7 | 979 | |||
AF540955.1 | - | 7 | 871 | |||
AK126803.1 | - | 10 | 1129 | |||
AK291823.1 | - | 7 | 941 | |||
AK296751.1 | - | 6 | 648 | |||
BC013238.1 | - | 8 | 943 | |||
BC024254.2 | - | 7 | 934 | 3531592 | ||
U87166.1 | - | 9 | 1036 |