Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015053.1 | NP_001015053.1 | 23 | 3193 | ||
NM_005474.4 | NP_005465.2 | 23 | 3190 | |||
XM_005256905.1 | XP_005256962.1 | 23 | 3188 | |||
XM_005256906.4 | XP_005256963.1 | 23 | 3149 | |||
XM_011524149.1 | XP_011522451.1 | 23 | 3164 | |||
XM_011524150.1 | XP_011522452.1 | 22 | 2940 | |||
XM_017023988.1 | XP_016879477.1 | 25 | 3302 | |||
XM_017023989.1 | XP_016879478.1 | 25 | 3268 | |||
XM_017023990.1 | XP_016879479.1 | 25 | 3293 | |||
XM_017023991.1 | XP_016879480.1 | 25 | 3299 | |||
XM_017023992.1 | XP_016879481.1 | 25 | 3288 | |||
XM_017023993.1 | XP_016879482.1 | 25 | 3275 | |||
XM_017023994.1 | XP_016879483.1 | 21 | 3256 | |||
XM_017023995.1 | XP_016879484.1 | 23 | 3158 | |||
XM_017023996.1 | XP_016879485.1 | 24 | 3365 | |||
GenBank mRNA | AB011172.2 | - | 21 | 2907 | ||
AB177853.1 | - | 13 | 1642 | |||
AF039691.1 | - | 18 | 2295 | |||
AF132608.1 | - | 22 | 2858 | |||
BC038423.1 | - | 12 | 1472 | |||
BC051824.1 | - | 23 | 3069 | 6043491 |