Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TROAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005480.3 | NP_005471.3 | 8 | 968 | ||
XM_006719181.1 | XP_006719244.1 | 8 | 912 | |||
XM_011537723.1 | XP_011536025.1 | 8 | 912 | |||
XM_011537724.1 | XP_011536026.1 | 7 | 708 | |||
GenBank mRNA | AK296811.1 | - | 8 | 855 | ||
AK299302.1 | - | 8 | 775 | |||
AK309716.1 | - | 5 | 389 | |||
AK310709.1 | - | 7 | 706 | |||
AK315211.1 | - | 8 | 912 | |||
BC032583.1 | - | 8 | 880 | 5498800 | ||
U04810.2 | - | 8 | 911 |