Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135729.1 | NP_001129201.1 | 15 | 2240 | ||
NM_001135730.1 | NP_001129202.1 | 14 | 2175 | |||
NM_001135732.1 | NP_001129204.1 | 15 | 2313 | |||
NM_005488.2 | NP_005479.1 | 15 | 2310 | |||
NR_024194.1 | 15 | 2339 | ||||
NR_024195.1 | 14 | 2260 | ||||
XM_011529818.2 | XP_011528120.1 | 16 | 2327 | |||
XM_011529820.2 | XP_011528122.1 | 14 | 2178 | |||
XM_017028529.1 | XP_016884018.1 | 16 | 2324 | |||
XM_017028530.1 | XP_016884019.1 | 14 | 2175 | |||
GenBank mRNA | AF040251.1 | - | 1 | 833 | ||
AJ006973.1 | - | 15 | 2245 | |||
AK026576.1 | - | 15 | 2263 | |||
AK090723.1 | - | 3 | 1819 | |||
AK091958.1 | - | 3 | 2501 | |||
AK097066.1 | - | 13 | 1821 | |||
AK097926.1 | - | 17 | 2526 | |||
AY358980.1 | - | 9 | 1243 | |||
BC040006.1 | - | 15 | 2203 | |||
BC046151.1 | - | 15 | 2191 | 6470390 |