Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005490.2 | NP_005481.2 | 10 | 2124 | ||
XM_005259467.1 | XP_005259524.1 | 9 | 2161 | |||
XM_005259472.2 | XP_005259529.1 | 8 | 1774 | |||
XM_006722602.1 | XP_006722665.1 | 9 | 1801 | |||
XM_011527608.2 | XP_011525910.1 | 8 | 2578 | |||
XM_011527609.2 | XP_011525911.1 | 8 | 2075 | |||
GenBank mRNA | AF124249.1 | - | 10 | 2081 | ||
AK225386.1 | - | 10 | 2047 | |||
AK290293.1 | - | 10 | 2200 | |||
AK292781.1 | - | 10 | 2047 | |||
AK299406.1 | - | 8 | 1784 | |||
AY358406.1 | - | 10 | 2081 | |||
BC006281.2 | - | 10 | 2061 | 3958342 |