Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 13 | 2055 | ||
XM_006714945.2 | XP_006715008.1 | 13 | 1535 | |||
XM_011514205.2 | XP_011512507.1 | 11 | 1803 | |||
XM_017010149.1 | XP_016865638.1 | 13 | 1443 | |||
GenBank mRNA | AB008515.1 | - | 13 | 1744 | ||
AB055311.1 | - | 13 | 2055 | |||
AF306510.1 | - | 13 | 1593 | |||
AK313334.1 | - | 12 | 1446 | |||
BC052781.1 | - | 13 | 2216 | |||
BC063849.1 | - | 11 | 1311 | 6186252 |